Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Syringe filters, Econofilters
Leverancier: Agilent
These syringe filters have a PP housing.
  • Fast and efficient filtration
  • Low level of extractables
  • High flow rate
  • Maximum sample loading capacity
login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat
Order
Expand All / Collapse All
PTFE
Poriëngrootte Diameter VPE VWR catalogus nummer Hoeveelheid
0,2 µm 25 mm 1.000 HEWL5190-5267
0,2 µm 13 mm 1.000 HEWL5190-5265
0,45 µm 13 mm 1.000 HEWL5190-5266
0,45 µm 25 mm 1.000 HEWL5190-5268
Regenerated cellulose
Poriëngrootte Diameter VPE VWR catalogus nummer Hoeveelheid
0,2 µm 15 mm 1.000 HEWL5190-5310
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Specificaties
∅ (mm) 25 mm 13 mm 25 mm 13 mm 25 mm 13 mm 25 mm 13 mm 15
Poriëngrootte 0,2 µm 0,45 µm 0,2 µm 0,45 µm 0,2 µm 0,45 µm 0,2 µm 0,45 µm 0,2 µm 0,45 µm 0,2 µm
Filtratieoppervlak (cm²) 2,98 0,92 2,98 0,92 2,98 0,92 2,98 0,92
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid