Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
HPLC columns, MonoChrom® C18
Leverancier: Agilent Technologies
MonoChrom® columns are ideal for separation of organic amines, organic acids, amphoteric compounds and neutral compounds. Advanced silica production and silane bonding techniques allow for the production of material with exceptional base-deactivated characteristics.
login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat
Order
Expand All / Collapse All
Kolomtype Fase Poriëngrootte Partikelgrootte Ø inw.xl VPE VWR catalogus nummer Hoeveelheid
Analytical C18 110 Å 3 µm 2,0×30 mm 1 VARIA0400030X020
Analytical C18 110 Å 3 µm 2,0×100 mm 1 VARIA0400100X020
Analytical C18 110 Å 3 µm 3,0×150 mm 1 VARIA0400150X030
Analytical C18 110 Å 3 µm 4,6×150 mm 1 VARIA0400150X046
Analytical C18 110 Å 5 µm 2,0×50 mm 1 VARIA0505050X020
Analytical C18 110 Å 5 µm 2,0×100 mm 1 VARIA0505100X020
Analytical C18 110 Å 5 µm 4,6×150 mm 1 VARIA0505150X046
Analytical C18 110 Å 5 µm 4,6×250 mm 1 VARIA0505250X046
Analytical C18 110 Å 10 µm 3,9×300 mm 1 VARIA0504300X039
Analytical C18 110 Å 10 µm 4,0×300 mm 1 VARIA0504300X040
Analytical C18 110 Å 10 µm 4,6×250 mm 1 VARIA0504250X046
Guard columns
Kolomtype Fase Poriëngrootte Partikelgrootte VPE VWR catalogus nummer Hoeveelheid
Guard Column C18 110 Å 3 µm 3 VARIA0400MG2
Guard Column C18 110 Å 5 µm 3 VARIA0505MG2
Guard Column C18 110 Å 5 µm 3 VARIA0505MG
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid