Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
    • In Stock (19)
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
Handcrèmes
SearchPresentationType-VERTICAL
SearchResultCount:"27"
  List view Grid view Easy View (new)
Sorteer op:
Leverancier:  Deb-STOKO
Omschrijving:   This intensive care (W/O) cream contains a ceramide complex and grapeseed oil. It is ideal for dry and sensitive skin and regenerates and deeply nurtu...
Leverancier:  Deb-STOKO
Omschrijving:   Stokolan® Light Gel is a vitalising, non greasy care gel with moisturising glycerol and urea. The gel is very easy to apply and is absorbed into the s...
Leverancier:  Deb-STOKO
Omschrijving:   Aqua PURE is a barrier cream (W/O/W emulsion) that protects the skin when working with aqueous or water-miscible industrial substances and performing ...
Leverancier:  Deb-STOKO
Omschrijving:   Hand cream, Stokolan®, Cream, Vochtinbrengend, Buis, Recipiëntinhoud (ml): 100
Leverancier:  Deb-STOKO
Omschrijving:   Versatile skin protection product (O/W emulsion) for sensitive skin exposed to changing and non specific stresses. Can also be used under gloves. The ...
Leverancier:  Deb-STOKO
Omschrijving:   Stokoderm®​ Triple Active Gel with Deb Active Defence Molecules (DADM) designed for application under occupational gloves to improve hand health and c...
Leverancier:  Deb-STOKO
Omschrijving:   O/W emulsion against water-insoluble workplace substances. It helps to stop resins clinging and protects from oils and greases. Suitable for use on ha...
Catalogus nummer: (115-2249)
Leverancier:  Deb-STOKO
Omschrijving:   O/W emulsion against water-insoluble workplace substances. It helps to stop resins clinging and protects from oils and greases. Suitable for use on ha...
UOM:  1 * 9 ST
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8  Of 27
  1  2  3  4  Volgende