U heeft gezocht op: Vloeibare en schuimende huiddesinfectiemiddelen
10  results were found
SearchResultCount:"10"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: GOJO Industries - Europe Ltd
Omschrijving: Spa-inspired foam hand wash offers luxurious lather and a pleasant botanical fragrance.
Veiligere gezondheid van mens en milieu
Leverancier: GOJO Industries - Europe Ltd
Omschrijving: A no-rinse hygienic hand rub that feels great to use and is clinically proven to maintain skin health.
Catalogus nummer: (GOJO9762-06-EEU)
Leverancier: GOJO Industries - Europe Ltd
Omschrijving: A no-rinse hygienic hand rub that feels great to use and is clinically proven to maintain skin health.
UOM: 1 * 6 ST
Catalogus nummer: (GOJO1904-02-EEU00)
Leverancier: GOJO Industries - Europe Ltd
Omschrijving: A hygienic hand sanitising foam formulation, proven to help maintain skin health.
UOM: 1 * 1 ST
Catalogus nummer: (GOJO5179-03-EEU00)
Leverancier: GOJO Industries - Europe Ltd
Omschrijving: Glad schuimende handzeep met breed spectrum kiemdodende kracht en hydraterende eigenschappen. Hypoallergeen en mild voor de huid voor frequent gebruik...
UOM: 1 * 3 ST
Leverancier: Deb-STOKO
Omschrijving: Hypoallergenic, alcohol-based foam sanitiser for use in food industry. Compatible for use with latex and nitrile gloves.
Leverancier: Deb-STOKO
Omschrijving: This is a solvent-free workshop cream with plastic microbeads, effective against strong and adherent soiling such as greases, oils and fuels.
Leverancier: Deb-STOKO
Omschrijving: This is a heavy duty workshop soap without microbeads, effective against strong and adherent soiling.
Leverancier: Diversey
Omschrijving: Desinfecteert de handen in één eenvoudige stap zonder dat er water nodig is. Niet geclassificeerde zeer effectieve alcoholvrije foam handgel met een v...
Duurzame materialen
Veiligere gezondheid van mens en milieu
Leverancier: Essity
Omschrijving: These hand disinfectants contains 80% denatured alcohol (w/w) and does not need to be rinsed. This is developed for regular use without sticky residue...
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 10 of 10
no targeter for Bottom