Cartridge filters, Optimizer® PR 16 and PR 40
Supplier: ENTEGRIS
Certificates
About this item
The UPE membrane of these filters eliminates pre-wetting, which optimises the full use of the membrane and ensures maximum flow.
- Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
- Removes soft gel particles
- Provides superior downstream cleanliness, low surfactant binding and spontaneous wettability
Membrane: UPE; Support, cage, core and endcaps: HDPE
Maximum operating temperature: 60 °C
Maximum forward differential pressure: 3,4 bar at 25 °C
Maximum reverse differential pressure: 2,7 bar at 25 °C
Articles may not be available in all countries. Please contact your local Avantor office.
Product Information
- Catalog #Supplier #TypePore sizeFiltration areaPkAvailabilityPrice
- 514-0739CWAZ16KL1PR 16 cartridge filter with Kalrez® O-ring, Ø×L: 67×52 mm0,05 µm1500 cm²1Log in for availability1 * 1 STSpecifications:
Specifications
Environmentally Preferable - 514-0738CWAG40KL1PR 40 cartridge filter with Kalrez® O-ring, Ø×L: 67×113 mm0,2 µm3500 cm²1Log in for availability1 * 1 STSpecifications:
Specifications
Environmentally Preferable