Microtoom lemmet, MX35
Leverancier: Epredia
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-2791EA 271 EUR
720-2791 SHAN3150734 SHAN3051835 SHAN3050822
Microtoom lemmet, MX35
Microtomen Messen en handgrepen
Choose from HP35, MB22, or MX35 microtome blades.
  • Blades are available in high- and low-profile configurations
The MB22’s steeper angle allows users to accurately cut thinner sections faster.
HP35 disposable microtome blades for routine applications with a high proportion of harder tissue.
High-performance MX35 Premier disposable low-profile microtome blades for sectioning hard tissues and high-quality thin sections. Premier Plus blades provide greater durability, superior sharpness for lower blade friction and longer blade life.