U heeft gezocht op: Wafertransportsystemen
65  results were found
SearchResultCount:"65"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: ENTEGRIS INC
Omschrijving: Entegris offers a wide selection of H 44 series 4" square trays, covers and accessories to meet your changing needs. H 20 series trays are available i...
Leverancier: ENTEGRIS INC
Omschrijving: PFA, natural.
Leverancier: ENTEGRIS INC
Omschrijving: Holds one mask or reticle.
Leverancier: ENTEGRIS INC
Omschrijving: Spring boxes, Holds twelve trays and one cover or thirteen trays, Polypropylene, natural
Leverancier: ENTEGRIS INC
Omschrijving: Entegris offers a wide selection of H 20 series 2" square trays, covers and accessories to meet your changing needs. H 20 series trays are available i...
Catalogus nummer: (ENTEH207000011415)
Leverancier: ENTEGRIS INC
Omschrijving: Wafer handling trays to safely transport and protect silicon wafers.
UOM: 1 * 25 ST
Leverancier: ENTEGRIS INC
Omschrijving: Wafer handling trays to safely transport and protect silicon wafers.
Leverancier: ENTEGRIS INC
Omschrijving: For shipping and storage applications.
Leverancier: ENTEGRIS INC
Omschrijving: Beschikbaar voor wafers van 1,0 inch, 1,5 inch, 2,0 inch en 2,5 inch. Op het deksel grote ruimte voor etiketten.
Leverancier: ENTEGRIS INC
Omschrijving: Ultrapak® series multiple wafer shipping boxes are made of Ultrapure polypropylene materials that provide a clean shipping environment and assure low ...
Catalogus nummer: (ENTE125-175-47C02)
Leverancier: ENTEGRIS INC
Omschrijving: This process cassette is molded from STAT-PRO® 3000 and durable enough to withstand temperatures up to 120 °C. This saves process time and reduces the...
UOM: 1 * 1 ST
Leverancier: ENTEGRIS INC
Omschrijving: Deze 100 mm wafer carriers werden ontworpen voor een optimale prestatie in geautomatiseerde handelingen of voor het verwerken van LCD's en andere klei...
Leverancier: ENTEGRIS INC
Omschrijving: Wafer handling, Envelopes for wafer storage boxes, Envelopes, antistatic, PVC
Leverancier: ENTEGRIS INC
Omschrijving: Wafershieldâ„¢ high purity PP
Leverancier: ENTEGRIS INC
Omschrijving: Multiple film frame shippers provide a safe and effective way to ship and store up to 25 wafers, or singulated die, mounted to film frames. Special ap...
Leverancier: ENTEGRIS INC
Omschrijving: Film frame shippers provide a convenient, safe and ergonomic means to ship and store wafers mounted on film frames. Film frame shippers are designed t...
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 65
no targeter for Bottom