U heeft gezocht op: Wafer dragers
9  results were found
SearchResultCount:"9"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: ENTEGRIS INC
Omschrijving: Entegris offers a wide selection of H 44 series 4" square trays, covers and accessories to meet your changing needs. H 20 series trays are available i...
Leverancier: ENTEGRIS INC
Omschrijving: Entegris offers a wide selection of H 20 series 2" square trays, covers and accessories to meet your changing needs. H 20 series trays are available i...
Catalogus nummer: (ENTEH207000011415)
Leverancier: ENTEGRIS INC
Omschrijving: Wafer handling trays to safely transport and protect silicon wafers.
UOM: 1 * 25 ST
Leverancier: ENTEGRIS INC
Omschrijving: Wafer handling trays to safely transport and protect silicon wafers.
Leverancier: ENTEGRIS INC
Omschrijving: Nest between H 44 trays and/or covers.
Catalogus nummer: (ENTEH204000011415)
Leverancier: ENTEGRIS INC
Omschrijving: Wafer handling trays to safely transport and protect silicon wafers.
UOM: 1 * 100 ST
Leverancier: ENTEGRIS INC
Omschrijving: Nest between H 20 trays and/or covers.
Catalogus nummer: (ENTEH205000011415)
Leverancier: ENTEGRIS INC
Omschrijving: To support the semiconductor industry's requirements for advanced 150 mm wafer handling, Entegris offers a variety of wafer shipping and handling solu...
UOM: 1 * 100 ST
Catalogus nummer: (ENTEH203000011415)
Leverancier: ENTEGRIS INC
Omschrijving: Wafer handling trays to safely transport and protect silicon wafers.
UOM: 1 * 50 ST
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 9 of 9
no targeter for Bottom