Overalls, PP, with hood
These disposable overalls are made from non woven PP. Ideal for non critical applications in industrial, clinical or food processing environments.
- With hood and zip closure
- Elasticated waist, cuffs and ankles
- Individually packed
Weight: 40 to 50 g/m²
Colour: White
- Catalog No:
- 113-8123
- HOIT113946
- Model:
- With hood
- With hood
- Garment application:
- General Purpose, Medical, Food
- General Purpose, Medical, Food
- ColourClothing SizePk
- Overalls with hood, 40 g/m²: HOIT109736
Specifications
ModelWith hoodEnvironmentally PreferableGenderGarment applicationGeneral Purpose, Medical, Food - Overalls with hood, 50 g/m²Overalls with hood, 50 g/m²HOIT113946WhiteXXL501 * 50 ST292,00 €: HOIT113946
Specifications
ModelWith hoodEnvironmentally PreferableGenderGarment applicationGeneral Purpose, Medical, Food