Water penetration tester, acc. to Karsten
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIMELI7500EA 356 EUR
SIMELI7500
Water penetration tester, acc. to Karsten
Waterpenetratietesters
This Karsten tube penetration test is a simple test for measuring the degree or water penetration into building materials such as concrete, stone and plaster. The test kit consists of a glass tube filled with water, bonded to the test material with plastiline. Water pressure is then exerted on the surface. A graduated scale indicates, over time, the amount or water penetrated into the surface.
  • Tube material: Glass
  • Suitable for vertical and horizontal surfaces
Bestelinformatie: The TQC Karsten Tube Penetration Test contains 3 tubes for horizontal surfaces, 3 tubes for vertical surfaces (or at choice any other combination of total 6 tubes), a water bottle, marker and putty to place the tubes leak-tight on the surface. Additional vertical or horizontal tubes are available on request.