Digital microscope
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIMELD6182EA 261 EUR
SIMELD6182
Digital microscope
This USB microscope can be connected to a PC in order to determine the structure of surfaces or other objects. It is designed to analyse coating failures, imperfections or treatment quality.
  • USB camera with the optical accuracy of a LED microscope
  • Can be used vertically on ferrous material thanks to three strong magnets
  • Integrated white LEDs adjustable in strength for clear view without reflection
Suitable for measuring, editing and making videos, as well as determining the length, width, height and angle to the objects.
Leveringsinformatie: Supplied with a magnetic holder and the software.