Software for Ultrospec™ 2100 pro
Leverancier: Biochrom
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Ultrospec™
SCLI80-2110-73EA 338 EUR
SCLI80-2110-73
Software for Ultrospec™ 2100 pro
Software Laboratoriumsoftware
The spreadsheet interface software provides the facility to export results directly to an Excel spreadsheet.
Scan comprising data such as absorbance or wavelength, may be picked up as columns of numbers and converted to a more conventional graph using the spreadsheet.
Results may then be formatted or manipulated as appropriate prior to inclusion in reports or archiving.