U heeft gezocht op: Wafer verzendcontainers
22  results were found
SearchResultCount:"22"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: ENTEGRIS INC
Omschrijving: Wafer handling, Wafer shippers, H 22 series, wafer size 76,2 mm (3"), Spring, low force, Materiaal: PP, Kleur: natuur
Leverancier: ENTEGRIS INC
Omschrijving: Beschikbaar voor wafers van 1,0 inch, 1,5 inch, 2,0 inch en 2,5 inch. Op het deksel grote ruimte voor etiketten.
Leverancier: ENTEGRIS INC
Omschrijving: This SmartStack® contactless horizontal wafer shippers safely protect and transport bumped wafers in full, thin or ultrathin thickness.
Catalogus nummer: (ENTEH6620-0009)
Leverancier: ENTEGRIS INC
Omschrijving: Wafer handling, Multiple wafer shipping boxes for 2,5" and smaller wafer handling, Assembly, shrink-wrapped, PP, natural
UOM: 1 * 80 ST
Leverancier: ENTEGRIS INC
Omschrijving: Ultrapak® series multiple wafer shipping boxes are made of Ultrapure polypropylene materials that provide a clean shipping environment and assure low ...
Leverancier: ENTEGRIS INC
Omschrijving: Multiple film frame shippers provide a safe and effective way to ship and store up to 25 wafers, or singulated die, mounted to film frames. Special ap...
Leverancier: ENTEGRIS INC
Omschrijving: H 22 Series
Conical pocket holds one wafer facedown contacting only the wafer’s edges.
Leverancier: ENTEGRIS INC
Omschrijving: Film frame shippers provide a convenient, safe and ergonomic means to ship and store wafers mounted on film frames. Film frame shippers are designed t...
Leverancier: ENTEGRIS INC
Omschrijving: Ultrapak® series multiple wafer shipping boxes are made of Ultrapure polypropylene materials that provide a clean shipping environment and assure low ...
Leverancier: ENTEGRIS INC
Omschrijving: Conical pocket holds one wafer face down contacting only the edge of the wafer.
Leverancier: ENTEGRIS INC
Omschrijving: Horizontal wafer shipper.
Sale
Leverancier: ENTEGRIS INC
Omschrijving: SmartStack® horizontal wafer shipping system is designed to reduce wafer breakage, die bond corrosion and particle contamination. These shipping syste...
Catalogus nummer: (ENTEHWS15010161C2M)
Leverancier: ENTEGRIS INC
Omschrijving: Horizontal wafer shipper
UOM: 1 * 24 ST
Leverancier: ENTEGRIS INC
Omschrijving: Ultrapak® series multiple wafer shipping boxes are made of Ultrapure polypropylene materials that provide a clean shipping environment and assure low ...
Catalogus nummer: (ENTEH6630-0009)
Leverancier: ENTEGRIS INC
Omschrijving: Wafer handling, Multiple wafer shipping boxes for 76,2 mm (3") wafer handling, H 6630 Series, Assembly, shrink-wrapped, PP, natural
UOM: 1 * 60 ST
Leverancier: ENTEGRIS INC
Omschrijving: For shipping and storage applications. Holds one wafer facedown contacting only the wafer’s edges.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 22
no targeter for Bottom