Electron micrography films, Kodak
Leverancier: Electron Microscopy Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KODK8010100EA 908 EUR
KODK8010100 730-3213 KODK1679273 KODK1493279 KODK8971160 KODK1662238 KODK1608124
Electron micrography films, Kodak
Autoradiografiefilms Folies
These micrography films offer considerable latitude in performance and flexibility for most electron microscopy applications. They are suitable for medium-energy electrons (40 to 100 ky) and acceptable results can also be obtained at higher electron levels.
  • Engineered to produce high-quality images
  • Transmission Electron Micrography (TEM) or electron diffraction studies
  • Versatile image capture in the transmission electron microscope for life and material research
Micrography Film 4489 with less curl and shorter pump-down times. Approximately half the speed of Image Film SO-163. Coated on a 0,18 mm Estar support, electron microscope film offers dimensional stability and eliminates the use of glass support.
Image Film SO-163 are twice faster with broad exposure and development ranges. Single film to collect many electrons with improvements in SNR or fewer electrons in exposure-limiting situations (when graininess is a reasonable trade-off for higher speed).
Opgelet: SO-163 film speed requires longer pump-down times and shows greater film curl. When using SO-163 for the first time, it is recommended to recalibrate the microscope exposure.