Salpeterzuur fuming ≥90.0% (by titrimetric analysis), BAKER ANALYZED® ACS, J.T.Baker®
Leverancier: Avantor
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
9624-05EA
710.38
EUR
9624-02
9624-05
Salpeterzuur fuming ≥90.0% (by titrimetric analysis), BAKER ANALYZED® ACS, J.T.Baker®
Salpeterzuur
Formule:
HNO₃ MW: 63,01 g/mol Kookpunt: 87 °C (1013 hPa) Smeltpunt: –42 °C Dichtheid: 1,5 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011349 CAS nummer: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 8,I Bereik: 01-2119487297-23 Merck Index: 12,06672 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (as HNO₃) (by acidimetry) | ≥90.0 % |
Dilution Test | Passes Test |
ACS - Residue after Ignition | ≤0.002 % |
ACS - Dissolved Oxides (as N₂O₃) | ≤0.1 % |
ACS - Chloride (Cl) | ≤0.7 ppm |
ACS - Sulfate (SO₄) | ≤5 ppm |
Trace Impurities - Arsenic (As) | ≤0.3 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤2.0 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...