Mini spray dryer B-290
Leverancier: Büchi
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BUCH044700EA 0 EUR
BUCH044700 BUCH044699
Mini spray dryer B-290
Sproeidrogers
Mini spray dryer B-290 is designed to visualise and trial the spray drying process for wide range of applications such as pharma, materials, chemistry, food, feed, beverages, etc. The Advanced models allow for working with acids, organic solvents or mixtures of aqueous- and organic solvents safely in combination with inert atmospheres and the following accessories; inert loop B-295 and dehumidifier B-296.
  • Integrated 2-fluid nozzle
  • The inner wall of the glass cyclone is coated with an electrically conductive layer to prevent any electrostatic charge of the product
  • The glassware is easy to assemble and dissassemble for cleaning
  • Operated with compressed air or nitrogen, 200 to 800 L/h, 5 to 8 bar
  • Average residence time: 1 - 1,5 s