SPE sorbents, LiChrolut® EN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.19853.0020EA 230 EUR
MERC1.19853.0020
SPE sorbents, LiChrolut® EN
LiChrolut® EN was especially developed for application in environmental analysis where highly contaminated samples occur and very polar organic compounds have to be analysed. In comparison to LiChrolut® RP-18, LiChrolut® EN has a tenfold higher capacity.
  • Use of common organic solvents, buffer solutions, acids and bases over the entire pH range
  • Saving of solvent, as little solvent is required for conditioning and elution of the cartridge bed
  • Time saving, as less adsorbent requires less time for conditioning and drying
  • Improved analysis, as the reduced quantity of solvent required for elution leads to a lower degree of contamination and to an increase in detection sensitivity