VWR® ARISTAR®, ICP Multi-Element Solution According USP 232 Parental dose, 6 Elements 10 mg/L in 15% Hydrochloric Acid
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR® ARISTAR®
85039.180EA 299 EUR
85039.180
VWR® ARISTAR®, ICP Multi-Element Solution According USP 232 Parental dose, 6 Elements 10 mg/L in 15% Hydrochloric Acid
Standaarden Kationische standaarden Standaarden met meerdere elementen
Six elements: 10 mg/L each of Ir; Pt; Os; Rh; Pd; Ru in HCl 15%.
Certificaten: USP 232. Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022