Bottle top tilt measures
Leverancier: Hach Lange
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HACH2220038EA 346.08 EUR
HACH2220038
Bottle top tilt measures
Flesdispenseropzet
Borosilicate glass, clear.
  • Complete with fixing spring
  • Cone with ground joint
This dispenser head is completed by a 1 L Erlenmeyer flasks for repetitive dispensing of the same volume. Inversion of unit fills measuring tube with 10 ml of liquid for easy dispensing.