Insulate containers, with locking clamps
Supplier: BOCHEM
Certificates
About this item
Stainless steel 18/10.
- With three locking clamps, lid and silicone gasket
- Valve in the lid compensates possible over and under pressure
- Very good chemical and temperature resistance
Product Information
- Catalog #Supplier #CapacityHeightInt.ØPkAvailabilityPrice
- Supplier Article Number: 8085Local Article Number: BOCH8085Specifications:
Specifications
Environmentally Preferable - Supplier Article Number: 8086Local Article Number: BOCH8086Specifications:
Specifications
Environmentally Preferable - Supplier Article Number: 8087Local Article Number: BOCH8087Specifications:
Specifications
Environmentally Preferable