Elbow Pad for Pipetting
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BHIT723103EA 72.2 EUR
BHIT723103
Elbow Pad for Pipetting
Ergonomische steunen en hulpmiddelen Elleboogsteunen
This elbow pad helps users to feel more comfortable while pipetting. The visco elastic material of the pad relieves contact stress, pain and discomfort underneath the elbow. Ideal for long pipetting series, work requiring high concentration (e.g. micro plate work) and for any work where people need a cushion underneath the elbow or wrist.
  • Compact size takes up little desk space
  • Improves pipetting ergonomics
  • Coating is pleasant to the skin
  • Easy to clean with washing up liquid or ethanol (70%)