TOC gas generators
Leverancier: Peak Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PEAK42-1030EUEA 0 EUR
PEAK42-1030EU PEAK40-1040 PEAK42-1050 PEAK42-1030UK
TOC gas generators
Gasgeneratoren Persluchtstations
TOC gas generators produce up to 1500 ml/min of carrier/combustion gas from in house compressed air supply. They are recommended for NPOC (Non-purgeable organic carbon). They have a CO₂ content of less than 1,0 ppm and a pressure dew point of −70 °C.
  • Few moving parts with minimum maintenance requirements
  • Flexibility to position generator where required, wall- mountable for small labs
  • Completely silent in operation
  • Maximum inlet pressure of 6,9 to 8,2 bar