Netherlands
Order Entry
Water sampling bottles, glass, narrow neck, Boston type, round
Supplier:  EP SCIENTIFIC
   Sustainable Options Available
Certificates
undefined
Water sampling bottles, glass, narrow neck, Boston type, round
Supplier:  EP SCIENTIFIC
   Sustainable Options Available
Specifications
  • Catalog No:
  • EPSC349-0500
  • EPSC329-0250
  • EPSC229-0500
  • EPSC349-0125
  • EPSC229-1000
  • EPSC329-1000
  • EPSC229-0125
  • EPSC229-0250
  • EPSC329-0500
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Clear
Product Information
  • Catalog #
    Supplier #
    Capacity
    Dosage
    Closure type
    Cap colour
    With
    Availability
    Price
  • Certified, for semi-VOA, pesticides, PCBs and metals, Ø×H: 60×136 mm
    Certified, for semi-VOA, pesticides, PCBs and metals, Ø×H: 60×136 mm
    EPSC329-0250
    329-0250
    250 ml
    Undosed
    Cap, PP
    White
    unattached analysis label
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 75,50 €
    75,50 €
    1 * 12 ST
       75,50 €
    Supplier Article Number: 329-0250
    Local Article Number: EPSC329-0250
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Certified, for semi-VOA, pesticides, PCBs and metals, Ø×H: 74×168 mm
    Certified, for semi-VOA, pesticides, PCBs and metals, Ø×H: 74×168 mm
    EPSC329-0500
    329-0500
    500 ml
    Undosed
    Cap, PP
    White
    unattached analysis label
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 104,00 €
    104,00 €
    1 * 12 ST
       104,00 €
    Supplier Article Number: 329-0500
    Local Article Number: EPSC329-0500
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Certified, for semi-VOA, pesticides, PCBs and metals, Ø×H: 93×286 mm
    Certified, for semi-VOA, pesticides, PCBs and metals, Ø×H: 93×286 mm
    EPSC329-1000
    329-1000
    950 ml
    Undosed
    Cap, PP
    White
    unattached analysis label
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 127,00 €
    127,00 €
    1 * 12 ST
       127,00 €
    Supplier Article Number: 329-1000
    Local Article Number: EPSC329-1000
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Processed, for semi-VOA, pesticides, PCBs and metals, Ø×H: 48×112 mm
    Processed, for semi-VOA, pesticides, PCBs and metals, Ø×H: 48×112 mm
    EPSC229-0125
    229-0125
    125 ml
    Undosed
    Cap, PP
    White
    unattached analysis label
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 81,20 €
    81,20 €
    1 * 12 ST
       81,20 €
    Supplier Article Number: 229-0125
    Local Article Number: EPSC229-0125
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Processed, for semi-VOA, pesticides, PCBs and metals, Ø×H: 74×168 mm
    Processed, for semi-VOA, pesticides, PCBs and metals, Ø×H: 74×168 mm
    EPSC229-0500
    229-0500
    500 ml
    Undosed
    Cap, PP
    White
    unattached analysis label
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 133,00 €
    133,00 €
    1 * 12 ST
       133,00 €
    Supplier Article Number: 229-0500
    Local Article Number: EPSC229-0500
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Processed, for semi-VOA, pesticides, PCBs and metals, Ø×H: 60×136 mm
    Processed, for semi-VOA, pesticides, PCBs and metals, Ø×H: 60×136 mm
    EPSC229-0250
    229-0250
    250 ml
    Undosed
    Cap, PP
    White
    unattached analysis label
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 83,90 €
    83,90 €
    1 * 12 ST
       83,90 €
    Supplier Article Number: 229-0250
    Local Article Number: EPSC229-0250
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Processed, for semi-VOA, pesticides, PCBs and metals, Ø×H: 93×286 mm
    Processed, for semi-VOA, pesticides, PCBs and metals, Ø×H: 93×286 mm
    EPSC229-1000
    229-1000
    950 ml
    Undosed
    Cap, PP
    White
    unattached analysis label
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 107,00 €
    107,00 €
    1 * 12 ST
       107,00 €
    Supplier Article Number: 229-1000
    Local Article Number: EPSC229-1000
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Amber
Product Information
  • Catalog #
    Supplier #
    Capacity
    Dosage
    Closure type
    Cap colour
    With
    Availability
    Price
  • Certified, for semi-VOA, pesticides, PCBs and metals, Ø×H: 76×168 mm
    Certified, for semi-VOA, pesticides, PCBs and metals, Ø×H: 76×168 mm
    EPSC349-0500
    349-0500
    500 ml
    Undosed
    Cap, PP
    White
    unattached analysis label
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 111,00 €
    111,00 €
    1 * 12 ST
       111,00 €
    Supplier Article Number: 349-0500
    Local Article Number: EPSC349-0500
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Certified, for semi-VOA, pesticides, PCBs and metals, Ø×H: 48×112 mm
    Certified, for semi-VOA, pesticides, PCBs and metals, Ø×H: 48×112 mm
    EPSC349-0125
    349-0125
    125 ml
    Undosed
    Cap, PP
    White
    unattached analysis label
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 77,50 €
    77,50 €
    1 * 12 ST
       77,50 €
    Supplier Article Number: 349-0125
    Local Article Number: EPSC349-0125
    Specifications:
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact