96-Well format pipetting device, Vaccu-Pette SP Bel-Art
Leverancier: Bel-Art
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
612-6020EA 131 EUR
612-6020 612-6021
96-Well format pipetting device, Vaccu-Pette SP Bel-Art
Microplaatdispensers
Vaccu-Pette 96-well pipetting device can be used to aspirate or dispense 96 wells in less than 10 seconds, speeding up repetitive washing, dispensing or aspirating of wells by working simultaneously.
  • Transparent plastic construction
  • Available in sterile/disposable or autoclavable/reusable models
  • Ideal for efficient washing, dispensing, or aspirating of wells
The device speeds up repetitive washing, dispensing or aspirating of wells by working simultaneously. It has a vacuum outlet at one end, and a short length of 3,2 mm internal diameter tubing is supplied for syringe connection.
Sterile/disposable model is moulded from clear, disposable plastic that can be used only once and cannot be autoclaved or gas sterilised. Autoclavable/reusable model, moulded from PC, must be sterilised by autoclaving at no more than 121 °C for 20 minutes at 15 psi prior to use in order to avoid contamination. It cannot be gas sterilised due to the possibility of residual trapped gases.
Simply connect a disposable plastic syringe and withdraw the plunger to transfer liquid. Excellent for simultaneously changing media or for the addition/removal of reagents from a 96-well plate.
Bestelinformatie: Requires a 30 ml or larger disposable sterile syringe, which must be ordered separately.
Leveringsinformatie: Complete with a 7,6 cm long piece of 3,2 mm tubing for syringe connection. The autoclavable, reusable polycarbonate pipetting device is supplied in a sealed pack with tubing enclosed.