Cleaning and immersion containers, PREMIUM Line
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MSGA18433EA 162 EUR
MSGA18433 MSGA18435 MSGA18434
Cleaning and immersion containers, PREMIUM Line
Made from sheet steel. Interior tin-coated, exterior with high quality powder coating.
  • Sturdy design
  • Items can be immersed in a optionally available part basket
  • Self-closing cover in the event of fire due to an integrated thermal release (fusible link)
  • High stability, safe working
Ideal for cleaning and immersion of small items.
Certificaten: Approved safety products (FM)