Anti-BCL11A Rabbit Polyclonal Antibody
AVIVARP39569_P050
- Antibody type:Primary
- Antigen name:B-cell CLL/lymphoma 11A (zinc finger protein)
- Antigen symbol:BCL11A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:53335
- Antigen synonyms:ZNF856|BCL11A-XL|CTIP1|BCL11a-M|HBFQTL5|BCL11A-L|EVI9|BCL11A-S
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: BCL11A (B-cell CLL/lymphoma 11A (zinc finger protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: