Anti-SRI Rabbit Polyclonal Antibody
Leverancier: Aviva Systems Biology
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AVIVARP62986_P050EA 371 EUR
AVIVARP62986_P050
Anti-SRI Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: SRI (sorcin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now