Anti-GPR157 Rabbit Polyclonal Antibody
Leverancier: Aviva Systems Biology
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AVIVARP68397_P050EA 361 EUR
AVIVARP68397_P050
Anti-GPR157 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: GPR157 (G protein-coupled receptor 157)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human