Anti-NARS2 Rabbit Polyclonal Antibody
Leverancier: Abgent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABGEAP7585BEA 348 EUR
ABGEAP7585B
Anti-NARS2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: NARS2 (asparaginyl-tRNA synthetase 2, mitochondrial (putative))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human