Anti-E2F1 Rabbit Polyclonal Antibody
Leverancier: Abgent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABGEAP7593CEA 348 EUR
ABGEAP7593C
Anti-E2F1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: E2F1 (E2F transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human