Anti-E2F1 Rabbit Polyclonal Antibody

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP7593DSEA 99 EUR
ABGEAP7593DS ABGEAP7593D
Anti-E2F1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: E2F1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR