Anti-CCNB1 Rabbit Polyclonal Antibody
Leverancier: Abgent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABGEAP7598BEA 348 EUR
ABGEAP7598B
Anti-CCNB1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: CCNB1 (cyclin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse