Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

Anti-LUM Rabbit Polyclonal Antibody

Leverancier: Abgent
Type: Primary
Antigen: LUM (lumican)
Clonality: polyclonal
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse

login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat


Expand All / Collapse All
Maat VPE VWR catalogus nummer Hoeveelheid
400 µl 400 µl ABGEAP7605A
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol LUM
Antigen name Lumican
Reactivity Human, Mouse Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype none
Western blot Yes
ImmunoChemistry Yes
Flow cytometry Yes
Antigen synonyms SLRR2D, LUM, Keratan sulfate proteoglycan lumican, LDC, KSPG lumican
Epitope N-terminal
Opslagtemperatuur Cool
Shipping temperature Ice


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid