Anti-EPHA7 Rabbit Polyclonal Antibody
Leverancier: Abgent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABGEAP7612BEA 348 EUR
ABGEAP7612B
Anti-EPHA7 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: EPHA7 (EPH receptor A7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human