Anti-EPHA10 Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP7614AEA 348 EUR
ABGEAP7614A
Anti-EPHA10 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: EPHA10 (EPH receptor A10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR