Anti-PCSK2 Rabbit Polyclonal Antibody
Leverancier: Abgent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABGEAP7617CEA 348 EUR
ABGEAP7617C
Anti-PCSK2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: PCSK2 (proprotein convertase subtilisin/kexin type 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human