Anti-EPHB6 Rabbit Polyclonal Antibody
Leverancier: Abgent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABGEAP7627AEA 348 EUR
ABGEAP7627A
Anti-EPHB6 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: EPHB6 (EPH receptor B6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human