Anti-DAZL Rabbit Polyclonal Antibody

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP7739DSEA 99 EUR
ABGEAP7739DS
Anti-DAZL Rabbit Polyclonal Antibody
Type: Primary
Antigen: DAZL (deleted in azoospermia-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR