Anti-PRKRA Rabbit Polyclonal Antibody
Leverancier: Abgent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABGEAP7744BEA 348 EUR
ABGEAP7744B
Anti-PRKRA Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: PRKRA (protein kinase, interferon-inducible double stranded RNA dependent activator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human