Anti-MRAS Rabbit Polyclonal Antibody

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP7748CSEA 99 EUR
ABGEAP7748CS
Anti-MRAS Rabbit Polyclonal Antibody
Type: Primary
Antigen: MRAS (muscle RAS oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR