Anti-KLF10 Rabbit Polyclonal Antibody
Leverancier: Abgent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABGEAP7749AEA 348 EUR
ABGEAP7749A
Anti-KLF10 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: KLF10 (Kruppel-like factor 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human