Anti-FKBP1A Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP7756CEA 348 EUR
ABGEAP7756C
Anti-FKBP1A Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: FKBP1A (FK506 binding protein 1A, 12kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR