Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

Anti-RHOA Rabbit Polyclonal Antibody

Leverancier: Abgent
Type: Primary
Antigen: RHOA (ras homolog family member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human



login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat
 

Order

Expand All / Collapse All
Maat VPE VWR catalogus nummer Hoeveelheid
80 µl 20 µG ABGEAP7757BS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Specificaties

Antigen symbol RHOA
Antigen name Ras Homolog Family Member A
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype none
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitope C-terminal
Opslagtemperatuur Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid