Anti-CAMKK1 Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP7768CEA 348 EUR
ABGEAP7768C
Anti-CAMKK1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: CAMKK1 (calcium/calmodulin-dependent protein kinase kinase 1, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR