Anti-LTBP1 Rabbit Polyclonal Antibody
Leverancier: Abgent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABGEAP7780BEA 348 EUR
ABGEAP7780B
Anti-LTBP1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: LTBP1 (latent transforming growth factor beta binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse