Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

Anti-CYP3A4 Rabbit Polyclonal Antibody

Leverancier: Abgent
Type: Primary
Antigen: CYP3A4 (cytochrome P450, family 3, subfamily A, polypeptide 4)
Clonality: polyclonal
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human

login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat


Expand All / Collapse All
Maat VPE VWR catalogus nummer Hoeveelheid
400 µl 400 µl ABGEAP7788C
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol CYP3A4
Antigen name cytochrome P450, family 3, subfamily A, polypeptide 4 Cytochrome P450, Family 3, Subfamily A, Polypeptide 4
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype none
Western blot Yes
Antigen synonyms Cytochrome P450 3A3, Taurochenodeoxycholate 6-alpha-hydroxylase, HLP, P450PCN1, CYPIIIA3, NF-25, HLp, P450-PCN1, CP33, Cytochrome P450 3A4, CYP3A3, CYPIIIA4, Quinine 3-monooxygenase, CYP3A, CP34, P450C3, Nifedipine oxidase
Epitope middle
Opslagtemperatuur Cool
Shipping temperature Ice


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid