Anti-CYP2W1 Rabbit Polyclonal Antibody

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP7792CSEA 99 EUR
ABGEAP7792CS
Anti-CYP2W1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: CYP2W1 (cytochrome P450, family 2, subfamily W, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR