Anti-DSG2 Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP7795AEA 348 EUR
ABGEAP7795A
Anti-DSG2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: DSG2 (desmoglein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR