Anti-FBLN2 Rabbit Polyclonal Antibody

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORIGTA307986EA 655 EUR
ORIGTA307986
Anti-FBLN2 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: FBLN2 (fibulin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR