Anti-TTC30A Rabbit Polyclonal Antibody

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORIGTA308001EA 655 EUR
ORIGTA308001
Anti-TTC30A Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: TTC30A (tetratricopeptide repeat domain 30A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR